Learn More
3M™ Disposable Protective Coverall 4520
$175.40 - $201.50
Specifications
Ankle Style Elastic
Collar Type Standard fold
Closure Type 2-way Zipper
Color White
Description 3 Layers of Melt-blown Polypropylene in-between 2 layers of Spunbound Polypropylene
View More Specs
Products 6
Catalog Number Mfr. No. Size Size (English) Chest Height (English) Size (English) Waist Price Quantity & Availability  
Catalog Number Mfr. No. Size Size (English) Chest Height (English) Size (English) Waist Price Quantity & Availability  
17-624-103
View Documents
3M™
7000088928
3X-Large 49 to 52 in. 76 to 78 in. 54 in.
Case of 25 for $194.80
Add to cart
17-624-104
View Documents
3M™
7000088929
4X-Large 52 to 55 in. 78 to 81 in. 55 in.
Case of 25 for $201.50
Add to cart
17-624-105
View Documents
3M™
7000088930
Large 39 to 43 in. 69 to 71 in. 51 in.
Case of 25 for $175.40
Add to cart
17-624-107
View Documents
3M™
7000088933
2X-Large 45 to 49 in. 73 to 76 in. 53 in.
Case of 25 for $175.40
Add to cart
17-624-106
View Documents
3M™
7000088931
Medium 36 to 39 in. 66 to 69 in. 50 in.
Case of 25 for $175.40
Add to cart
17-624-108
View Documents
3M™
7000088934
X-Large 43 to 45 in. 70 to 74 in. 52 in.
Case of 25 for $175.40
Add to cart
Description
Description
3M™ Protective Coveralls are designed for use in industrial workplaces as a physical barrier to help prevent skin contact from liquid splashes and dust in certain work environments. Seamless shoulder and sleeve tops help provide increased comfort and fewer potential entry points for contaminants. In addition, these coveralls meet ANSI 101-1996 size guidelines. 3M™ Disposable Protective Coverall Safety Work Wear 4520CS meets the following standards: Protective Clothing - standard EN 340, Type 6 Splash - EN 13034/(EN ISO 17491-4), Type 5 Dust - EN ISO 13982-1, Anti-Static* - EN 11492** and Nuclear - EN 1073 - 2. * Consult class results in Technical Data Sheet or User Instructions for more information** All apparel must be grounded for anti-static treatment to be effective
  • Protective coverall helps protect against certain light liquid splashes and hazardous dusts
  • Microporous PE laminate offers a barrier against hazardous particles and liquid splashes
  • Breathable, SMMMS fabric back panel helps improve air ventilation and reduce heat build-up
  • Reinforcement gusset between the legs to help provide increased durability during bending and squatting
  • Meets government standards for Type 6 Splashes, Type 5 Dust, Anti-Static and Nuclear protection
  • Elastic waist and ankles for convenience and freedom of movement
  • Two-way zipper with sealable storm flap for convenience and to help provide additional protection
  • 3M Catalog# 4520CS-BLK-3XL, 4520CS-BLK-4XL, 4520CS-BLK-L, 4520CS-BLK-M, 4520CS-BLK-XL, 4520CS-BLK-XXL
Specifications
Specifications
Elastic
2-way Zipper
3 Layers of Melt-blown Polypropylene in-between 2 layers of Spunbound Polypropylene
Breathable SMS
Yes
No
Standard fold
White
Knit
Serged, Bound
Construction, mining, oil and gas, Powder handling, abatement and remediation, cleaning and maintenance, provide basic barrier protection against light liquid splashes and hazardous dusts