3M™ Disposable Protective Coverall 4530

Manufacturer: 3M™ 4530CSBLKXXL

 View more versions of this product

Catalog No. 17-624-118

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance EN 13034, EN 13982, EN 1149, EN 1073, ISO 14116
    Material Breathable SMS
    Seam Type Serged and Bound
    Color Blue
    Size 2XL
    Coating Anti-static
    Disposable Yes
    Height (Metric) 186 to 194cm
    For Use With (Application) Construction, mining, oil and gas, Powder handling, abatement and remediation, cleaning and maintenance, provide basic barrier protection against light liquid splashes and hazardous dusts
    Latex Free No
    Collar Type Standard
    Closure Type Two-way zipper
    Size (English) Inseam 34 in.
    Size (Metric) Waist 134cm
    Construction Suit: SMS Polypropylene, Back panel: SMS Polypropylene, Zipper: Nylon on Polyester Braid, Elastic: Neoprene Rubber, Cuffs: Polyester, Thread: Polyester
    Ankle Style Elastic
    Size (English) Chest 45 to 49 in.
    Size (Metric) Chest 115 to 124cm
    Height (English) 73 to 76 in.
    Size (English) Waist 53 in.
    Cuff Style Knit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit