Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Disposable Protective Coverall 4540+
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Size (English) Chest:
36 to 39 in.
39 to 43 in.
43 to 45 in.
45 to 49 in.
49 to 52 in.
52 to 55 in.
Height (English):
66 to 69 in.
69 to 71 in.
70 to 74 in.
73 to 76 in.
76 to 78 in.
78 to 81 in.
Size (English) Inseam:
30 in.
32 in.
33 in.
34 in.
35 in.
36 in.
Description
3M™ Protective Coveralls are designed for use in industrial workplaces as a physical barrier to help prevent skin contact from liquid splashes and dust in certain work environments. Seamless shoulder and sleeve tops help provide increased comfort and fewer potential entry points for contaminants. In addition, these coveralls meet ANSI 101-1996 size guidelines.
- Protective coverall helps protect against certain light liquid splashes and hazardous dusts
- Lightweight, breathable material helps keep workers cool and comfortable
- Extra room in arms and legs allow for free movement on the job
- Seamless shoulders and sleeve tops help provide increased comfort and fewer potential entry points for contaminants
- Meets government standards for Type 6 Splashes, Type 5 Dust, Anti-Static and Nuclear protection
- Three panel hood design helps provide a more comfortable fit and a sleek, low-profile style.
- Does not contain components made from natural rubber latex to help prevent reactions from users with latex sensitivities or restrictions.
- 3M Catalog# 4540+BLK-3XL, 4540+BLK-4XL, 4540+BLK-L, 4540+BLK-M, 4540+BLK-XL, 4540+BLK-XXL
Specifications
Size | 2X-Large |
Material | Breathable SMS |
Color | White |
Disposable | Single-use |
Waist Style | Elastic |
Cuff Style | Knit |
Neck Style | Collared |
Closure Type | 2-way Zipper |
Seam Type | Serged, Bound |
Coating | Anti-static |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction