Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Disposable Protective Coveralls

Provide basic barrier protection against light liquid splashes and hazardous dusts

$160.10 - $247.09

Specifications

Ankle Style Elastic
Certifications/Compliance CE, EN 1149-1:2006, EN ISO 13982-1 Type 5, EN 13034 Type 6
Coating Anti-static
Closure Type 2-way Zipper with storm flap
Color White
View More Specs

Alert:

Before use wearer must read and understand the User Instructions provided as part of the product packaging. Failure to follow all instructions on the use of these protective clothing products and/or failure to wear the complete product during all periods of exposure may adversely affect the wearer's health, or lead to severe or life threatening illness or permanent disability. Final determination as to the suitability of these products for a particular use is the employer's responsibility.


Products 3
Catalog Number Mfr. No. Size Height (Metric) Size (Metric) Inseam Size (Metric) Waist Price Quantity & Availability  
Catalog Number Mfr. No. Size Height (Metric) Size (Metric) Inseam Size (Metric) Waist Price Quantity & Availability  
19-164-137
View Documents
3M™
7000088923
4X-Large 194 to 200 cm 91.44 cm 139.7 cm Case for $246.12
Add to cart
 
19-164-138
View Documents
3M™
7000088924
Large 200 to 206 cm 81.28 cm 129.54 cm Case for $160.10
Add to cart
 
19-164-136
View Documents
3M™
7000088922
3X-Large 186 to 194 cm 88.9 cm 137.16 cm Case for $247.09
Add to cart
 
Description

Description

  • Made of high quality microporous-laminated material
  • Elastic waist, ankles, and wrists for convenience and free movement
  • Two-way zipper with storm flap for convenience and extra protection
  • Low-linting properties and anti-static coating on both sides helps minimize static build-up and prevent the risk of static discharge while using
  • Applications:

    • Asbestos Abatement and Inspection
    • Exposure to Radioactive Particles
    • Lead Abatement
    • Mold Remediation/Mold Spores
    • Fertilizer Application (Dry/Granular)
    • Pesticide Application (Liquid)
    • General Industrial Clean-up and Processing
    • Grease (General)
    • Light-duty Building Cleaning
    • Machine or Vehicle Maintenance
    • Coal Dust
    • Fiberglass/Cellulose Insulation Installation
    • Food Processing
    • General Powder Handling
    • Non-hazardous Aerosols (sprays)
    • Non-hazardous Liquids
    • Paint/Coating Spraying
    • Paint/Coating Spraying (with Isocyanate paints)
    • Clean Room Applications (ISO Class 4 - 9)
    • Pharmaceutical Powder Handling
    • Metal particulate (chromium beryllium, etc.)
    • Metal Polishing
    • 3M Catalog# 4510CS-BLK-3XL , 3M Catalog# 4510CS-BLK-4XL , 3M Catalog# 4510CS-BLK-L

    Certifications

    European Standards

Specifications

Specifications

Elastic
Anti-static
White
Elastic
Bound
Asbestos inspection, Coal dust in power plants, Metal polishing, light-duty building cleaning, machine or vehicle maintenance, paint spraying, pharmaceutical, general industrial clean-up, insulation installation, general powder handling and food processing
CE, EN 1149-1:2006, EN ISO 13982-1 Type 5, EN 13034 Type 6
2-way Zipper with storm flap
White SMMMS Polypropylene laminated suit, Nylon on Polyester braid zipper, Neoprene Rubber elastic, Polyester thread
Polypropylene
Yes
Elastic
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit