Learn More
3M™ E·A·R·™ Caboflex™ Model 600 Hearing Protector
Ideal for intermittent hearing protection needs
$183.92 - $576.18
Specifications
Color Blue/yellow
Size One size fits to all
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Material Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Material Price Quantity & Availability  
19-049092
View Documents
3M™
7000002385
Replacement Pods Silicone/PVC Case of 500 for $576.18
Add to cart
17-380-8
View Documents
3M™
7000002297
Hearing Protector PVC/Silicone/ABS Pack of 10 for $183.92
Case of 10 PK for $963.50
Add to cart
Description
Description
Features soft, conical tips that swivel for easier fitting and greater comfort.
  • Silicone tips contain a foam core for an effective seal and excellent noise reduction
  • Flexible band can be worn under the chin for use with hard hats
  • Noise Reduction Rating 20dB when the band is worn under the chin; 17dB when the band is worn behind the head
  • CSA Class BL
  • 3M Catalog# 320-2001 , 3M Catalog# 320-2004
Specifications
Specifications
Blue/yellow
One size fits to all