Learn More
3M™ E·A·R™ Classic™ Ear Plugs
NRR: 29dB
Manufacturer:  3M™ 3121201
 View more versions of this product
Catalog No. 17-380-1
  Check Availability
Add to cart
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.
Specifications Description & Specifications
Specifications
Regular
Yes
Assembly, blasting, cleaning, demolition, electrical, facility maintenance, grinding, machine operations, painting, sanding, welding
Roll down foam
Aircraft, ships, vehicles
Uncorded, Packed in polybags
CSA Class AL
Yellow
Yes
Classic™ Earplug
PVC
Cylindrical
29dB