Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ E•A•R•™ Model 4000 Ear Muff

Catalog Number 19145470
Click to view available options
Model 4000 Ear Muff
The 3M products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
The 3M products must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.

Offers exceptional protection at an economical price

Lightweight earfmuffs provide excellent noise attenuation (Noise Reduction Rating: 27dB) with numerous comfort/convenience features.

  • Large earcup openings fit most ear sizes
  • Soft, wide cushions ensure even distribution of pressure
  • Vertical adjustment system on band allows quick and easy fitting

Color Black, Blue
Includes Ear plugs, ear muffs and banded hearing protectors, soft wide cushions, vertical adjustment system on band
Certifications/Compliance OSHA
Size Universal
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
Noise Reduction Rating (NRR) 27 dB
Product Type Ear Muffs
Material ABS
Style Multi-position
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit