3M™ E·A·R™ UltraFit™ Ear Plugs Get IT Item

NRR: 25dB

Manufacturer: 3M™ 3404004

 View more versions of this product

Catalog No. 17-380-5

  Check Availability
Add to Cart

Alert

The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.


    Specifications

    Product Type Ultrafit™ Earplug
    Hearing Protection Style Pre-molded/reusable
    Material Elastomeric Polymer
    Mission Type Aircraft, ships, vehicles
    Shape Multi-flange
    Style Corded
    Noise Reduction Range (NRR) 25dB
    Certifications/Compliance CSA Class AL
    Size One size fits to all
    Color Yellow
    Cord Material Vinyl
    Dielectric Yes
    Disposable No
    For Use With (Application) Triple flange design and soft Polymer construction for safe use and nonirritating protection

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit