Learn More
3M™ E·A·R™ UltraFit Plus™ Ear Plugs
NRR: 26dB
Manufacturer:  3M™ 3406004
 View more versions of this product
Catalog No. 19-042-401
  Check Availability
Add to cart
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Specifications Description & Specifications
Specifications
One size fits to all
Yes
Patented flange design for most comfortable fit of any reusable plug
Pre-molded/reusable
Multi-flange
26dB
Yellow
No
Ultrafit Plus™ Earplug
Elastomeric Polymer
Corded, without case
CSA Class AL