Learn More
3M™ Empore™ 7mm dia./3mL vol. Extraction Disk Cartridges
Provide a unique alternation to packed column technology
Manufacturer:  3M™ 4214SD
 View more versions of this product
Catalog No. 14-378-100
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Silica
Solid Phase extraction of analytes from complex sample matrices
Extraction disk cartridge
C8-SD
3mL
0.7cm
Silica
Polypropylene
0.1 oz.