3M™ Empore™ 96-Well Extraction Disk Plates
High-performance extraction disk plates for high-throughput SPE
Manufacturer: 3M™ 6065
 View more versions of this product
Catalog No. 14-378-59
  Check Availability
Add to Cart
    Specifications
    Product Type 96-Well extraction disk plate
    For Use With (Application) LC/MS, solid Phase extraction of analytes from complex sample matrices, for volumes of 1.2mL or less
    Material Polypropylene
    Resin Composition Universal
    Sorbent C2, C8, or C18 Silica
    Volume (English) Well 0.04 oz.
    Volume (Metric) Well 1.2mL
  • Description & Specifications
Related Products
Product Content Feedback