Learn More
3M™ Empore™ Anion and Cation Exchange-SR Extraction Disks
Provide significantly faster extraction time with no negative effect on recoveries
Manufacturer:  3M™ 2252
 View more versions of this product
Catalog No. 14-386-6
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Provide significantly faster extraction time with no negative effect on recoveries
Specifications
Specifications
EPA 549.1 and 552.1
Analytical Preparation of Aqueous Environmental Samples
90% AnIon Exchange
4.7 cm
Extraction disk
SAX