Learn More
3M™ Empore™ C8 and C18 Extraction Disks
Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples
Manufacturer:  3M™ 2315
 View more versions of this product
Catalog No. 14-386-3
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples
  • Effective for extracting PCBs, PNAs, organochlorine pesticides, phenoxy acid herbicides, phthalates, and triazines
  • Provide up to 90% solvent reduction, excellent processing speed, and contamination-free extractions with accurate and reproducible results
  • Available with a variety of packings in 47 and 90mm sizes
  • C8 disk is approved for use in EPA Method 549.1
  • C18 disks are approved for use in EPA Methods 506, 550.1, 525.2, 1613B, 508.1, and 3535; 14-386-3 is also approved for use in EPA Method 608
Specifications
Specifications
EPA 506, 550.1, 525.2, 1613 B, 508.1, and 3535
For Extracting PCBs, PNAs, Organochlorine Pesticides, Phenoxy Acid Herbicides, Phthalates, and Triazines
90%
9 cm
Extraction disk
C18