Learn More
3M™ Empore™ C8 and C18 Extraction Disks
Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples
Supplier:  3M™ 2215
 View more versions of this product
Catalog No. 14-386-2
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples
  • Effective for extracting PCBs, PNAs, organochlorine pesticides, phenoxy acid herbicides, phthalates, and triazines
  • Provide up to 90% solvent reduction, excellent processing speed, and contamination-free extractions with accurate and reproducible results
  • Available with a variety of packings in 47 and 90mm sizes
  • C8 disk is approved for use in EPA Method 549.1
  • C18 disks are approved for use in EPA Methods 506, 550.1, 525.2, 1613B, 508.1, and 3535; 14-386-3 is also approved for use in EPA Method 608
Specifications
Specifications
Silica
Extracting PCBs, PNAs, Organochlorine Pesticides, Phenoxy Acid Herbicides, Phthalates, and Triazines
Extraction Disk
4.7 cm
EPA 506, 550.1, 525.2, 1613 B, 508.1, and 3535
C18
90%
Product Suggestions
Product Suggestions