Learn More
3M™ Empore™ C8 and C18 Extraction Disks
Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples
Manufacturer:  3M™ 2214
 View more versions of this product
Catalog No. 14-386
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples
Specifications
Specifications
Silica
4.7 cm
Extraction disk
C8
EPA 506, 550.1, 525.2, 1613 B, 508.1, and 3535
For Extracting PCBs, PNAs, Organochlorine Pesticides, Phenoxy Acid Herbicides, Phthalates, and Triazines
90%