3M™ Empore™ Carbon Extraction Disks
Provides all the advantages of activated carbon in a compact and thin disk
Manufacturer: 3M™ 2272
 View more versions of this product
Catalog No. 14-378-062
  Check Availability
Add to Cart
    Specifications
    Product Type Activated Carbon extraction disk
    Diameter (Metric) Outer 4.7cm
    For Use With (Application) Extraction of volatile, highly Polar, and water soluble compounds from aqueous solutions
  • Description & Specifications
Related Products
Product Content Feedback