3M™ Empore™ Oil and Grease Extraction Disks
Reduce extraction times to an average of eight minutes/liter for most sample matrices
Manufacturer: 3M™ 2270
 View more versions of this product
Catalog No. 14-638-30A
  Check Availability
Add to Cart
    Specifications
    Product Type Oil and grease extraction disk
    Diameter (Metric) Outer 4.7cm
    For Use With (Application) For use in EPA Method 1664
    Includes Hydrophobic matrix
    Sorbent C18
  • Description & Specifications
Related Products
Product Content Feedback