Learn More
3M™ Empore™ SDB-XC Extraction Disks
Reduce extraction time and solvent use for analytical preparation of aqueous environmental samples
Manufacturer:  3M™ 2340
 View more versions of this product
Catalog No. 14-386-5
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Approved for use in EPA Method 515.2
  • Provide the next step in environmental sample preparation, soluble polar compound extraction; ideal for phenol compounds having poor recoveries by almost any known extraction technique
  • Provide up to 90% solvent reduction, excellent processing speed, and contamination-free extractions with accurate and reproducible results
  • Available with a variety of packings in 47 and 90mm sizes
Specifications
Specifications
Silica
Reversed
Solid Phase Extraction
SDB-XC
EPA 515.2
9 cm
Extraction disk