3M™ Fectoggles™ Eyewear
Performance of an impact goggle with the comfort of a spectacle
Manufacturer: 3M™ 164080000010
 View more versions of this product
Catalog No. 19-039-159
  Check Availability
Add to Cart
Alert
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.
    Specifications
    Includes Adjustable temple frame
    Lens Tint Clear
    Material Polycarbonate
    Size Medium
    Surface Coating DX Antifog hardcoat
    Lens Coating DX Antifog Hardcoat
    Type Safety Goggles
    Certifications/Compliance ANSI Z87.1-2010-High Impact; CSA Z94.3-2007 standards
  • Description & Specifications
Related Products
Product Content Feedback