Learn More
3M™ Fectoggles™ Eyewear
Performance of an impact goggle with the comfort of a spectacle
Manufacturer:  3M™ 7010045743
 View more versions of this product
Catalog No. 19-807-989
Add to cart
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.
Description
Description
  • Polycarbonate lens absorbs 99.9% UV
  • DX™ antifog hardcoat protects lens from fog, scratching and most chemicals
  • Unique lens has no parting lines to obstruct or distract wearer's view
  • Lightweight, effective eye protection for high-particulate areas
  • Contoured to conform to the shape of the face to optimize coverage
  • Foam flange seals the entire perimeter of the lens to keep out fine particulates and control sweat buildup
  • Fire-retardant, braided, elastic headband for snug fit
  • 3M Catalog# 16420-00000-10, 16408-00000-10, 16400-00000-10, 16412-00000-10
Compliance
Meets ANSI™ Z87.1-2010 and CSA Z94.3-2007 standards.
Specifications
Specifications
ANSI Z87.1-2010-High Impact; CSA Z94.3-2007 standards
DX Anti-fog Hardcoat
Large
Anti-fog, Scratch-resistant
Safety
Adjustable temple frame
Polycarbonate
Clear
Safety goggles