Learn More
3M™ Fectoids™ Eyewear
Adjustable lens angle
Specifications
Certifications/Compliance Meet ANSI Z87.1-2003 standards.
View More Specs
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulatinos and the user instructions, warnings, and limitations according to each product.
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
18-999-2522
View Documents
3M™
161010000020
N/A Lens Tint: Clear; Frame Color: Blue; Large This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
18-999-2744
Accessories View Documents
3M™
16205
N/A Lens Tint: Gray; Frame Color: Blue This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
  • Sports styling closely follows contours of the face to provide complete coverage
  • DX™ anti-fog hard coat protects lens from fog, scratching, and most chemicals
  • Lenses absorb 99.9% UV
  • Lens angle can be adjusted to one of three positions
  • Length of the temples can be adjusted to four settings for a custom fit
  • Lenses come in two sizes and can be changed quickly and easily without tools
Compliance
Meet ANSI Z87.1-2003 standards.
Specifications
Specifications
Meet ANSI Z87.1-2003 standards.