3M™ GVP-Series Belt-Mounted PAPR Assemblies
Designed to provide maximum comfort
Manufacturer: 3M™ GVP-1UNIMH
 View more versions of this product
Catalog No. 19-813-751
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Product Type Web Belt
    For Use With (Equipment) Parp respirator
    Filter Type HE Filter
    For Use With (Application) Assembly, facility maintenance, machine operation, painting, sanding
    Includes Motor blower, web belt, nimh battery, charger, flow meter, power cord, blower plugs
    Mounting Type Belt
    Certifications OSHA
    Offer high airflow to the worker.
    • Single replaceable cartridge system
    • Long-life motor
    • Batteries provides eight hours of service per charge. and have a light to indicate when to recharge
    • Contours to the hip and body for comfort
    To assemble a complete system, select from the following pieces:
    • Headgear, helmet, facepiece, head cover or hood
    • Compatible breathing tube
    • Belt-mounted PAPR assembly
    • Appropriate filters/cartridges for your application
    These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
  • Description & Specifications
Related Products
Product Content Feedback