Learn More
3M™ GVP-Series Belt-Mounted PAPR Assemblies
Designed to provide maximum comfort
Manufacturer:  3M™ GVP1U
 View more versions of this product
Catalog No. 18-039-700
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Alert:
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Includes:
  • Web belt or comfort belt, motor blower, battery, charger, flow meter, power cord, blower plugs
  • Comfort belt-mounted assembly includes above items but features padded CB-1000 comfort belt that adjusts from 26 to 54 in. (66 to 134cm)
Description
Description
Offer high airflow to the worker.
  • Single replaceable cartridge system
  • Long-life motor
  • Batteries provides eight hours of service per charge. and have a light to indicate when to recharge
  • Contours to the hip and body for comfort
To assemble a complete system, select from the following pieces:
  • Headgear, helmet, facepiece, head cover or hood
  • Compatible breathing tube
  • Belt-mounted PAPR assembly
  • Appropriate filters/cartridges for your application
These safety products must be used in accordance with OSHA regulations and the user instructions, warnings and limitations accompanying each product. Misuse may result in sickness or death.
Specifications
Specifications
OSHA
Parp respirator
Assembly, machine operation
Belt
Urethane Coated Belt
HE Filter
Motor blower, battery, charger, flow meter, power cord, blower plugs