3M™ H-800 Series Full Brim Vented Hard Hats

Manufacturer: 3M™ H811V

 View more versions of this product

Catalog No. 17-623-727

  Check Availability
Add to Cart

    Specifications

    Harness Style Full Brim
    Includes 4-point Ratchet Suspension and Standard Brow Pad
    Shelf Life 5 Year(s)
    Size (Metric) 168 to 197mm
    Certifications/Compliance ANSI/ISEA Z89.1-2009 Type I, Class C
    Color Tan
    Material HDPE
    Quantity 20 Case
    Suspension Type 4-point Ratchet
    Ventilated Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit