3M™ H-Series White Hood Assemblies

Used on 3M™ Belt-Mounted Powered Air Purifying Respirator (PAPR)s and Supplied Air Respirator Systems

Manufacturer: 3M™ H-421

 View all options for this product

Catalog No. 19-088-1270


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Color White
    For Use With (Equipment) Belt mounted powered air purifying respirator and supplied air respirator system
    Product Type Hood Assembly
    Includes Inner shroud and cap suspension
    Material Tychem™ QC Fabric
    Size Standard
    Certifications NIOSH

    • Feature lightweight Tychem™ QC Fabric
    • Comfortable headgear
    • Can be worn with facial hair and glasses
    • White

    3M H-400 Series Hoods

    • Feature lightweight Tychem™ QC Fabric and provide high levels of airflow for maximum comfort For pharmaceutical, laboratory, and health care applications as well as during painting and sanding operations H-400 is available with hardhat and choice of inner shroud or collar

    These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit