Learn More
3M™ Hard Hat
This headgear integrates hardhat, face shield, welding shield and respirator when used as directed.
Supplier:  3M™ 7000127456
Catalog No. 19-825-057
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
This headgear can be used with 3M™ GVP-Series, or 3M™ Adflo-Series Powered Air Purifying Respirators (PAPRs), or 3M™ Supplied Air Respirator Systems. This headgear meets the test requirements for Industrial Protective Eyewear (ANSI Z87.1-2003), and meets the test requirements for Industrial Head Protection Type II, Class G (ANSI Z89.1-2003). It has an assigned protection factor of 25. 3M Catalog# L-705SG-F
Specifications
Specifications
13.4 in.
L-705SG-F
1/Cs.