3M™ High-Productivity Floor Stripping Pads

Remove old floor finish, dirt build-up

Manufacturer: 3M™ 08278

 View more versions of this product

Catalog No. 19-039-903

  Check Availability
Add to Cart

    Specifications

    Color Black
    Diameter (English) 20 in.
    Diameter (Metric) 50.8cm
    For Use With (Equipment) Rotary or automatic machine
    Material Nylon
    Thickness (English) 0.5 in.
    For Use With (Application) Stripping
    Product Type High productivity floor pad

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit