Learn More
3M™ Tychem™ Hoods and Head Covers
Help protect against airborne particulates that have a concentration of up to 25 x PEL using head cover, or 1000 x PEL using hood
Manufacturer:  3M™ BE10L3
 View more versions of this product
Catalog No. 19-039-711
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
White
Polycoated Hood
Disposable polycoated tychem™ hood, wide-view acetate visor
Large
3.2 oz.
Acetate
3M™ air-mate™ and breathe easy™ powered air purifying respirator system and be series supplied air system
Protect against airborne particulates that have a concentration of up to 25 x pel using head cover, or 1000 x pel using hood
Tychem™ QC Fabric
0.1kg
OSHA
Sewn in elastic headband
Accessories