3M™ Tychem™ Hoods and Head Covers
Help protect against airborne particulates that have a concentration of up to 25 x PEL using head cover, or 1000 x PEL using hood
Manufacturer: 3M™ BE1250
 View more versions of this product
Catalog No. 18-778-100
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) 3M™ air-mate™ and breathe easy™ powered air purifying respirator system,3M™ air-mate™ and breathe easy™ powered air purifying respirator system
    Product Type Headcover
    For Use With (Application) Protect against airborne particulates that have a concentration of up to 25 x pel using head cover, or 1000 x pel using hood
    Includes Wide view acetate visor
    Material Tychem™ QC Fabric
    Size Regular
    Certifications OSHA
    Material (Lens) Acetate
    Color White
    Suspension Type Integrated
  • Description & Specifications
Related Products
Product Content Feedback