3M™ KX Eyewear
Offers visitors excellent eye protection and a sleek, contoured style
Manufacturer: 3M™ 121550000020
 View more versions of this product
Catalog No. 19-121-303
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
Alert
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
    Specifications
    Lens Coating Anti-fog coating
    Quantity 20/cs.
    Certifications/Compliance KX Visitor Spectacles meet the requirements of CSA Z94.3-2007 and the high impact requirements of ANSI Z87.1-2010.
    Lens Material Antistatic Duralite Polycarbonate
    Lens Tint Clear
  • Description & Specifications
Related Products
Product Content Feedback