3M™ L-Series Headgear Accessories

Manufacturer: 3M™ L14010

 View more versions of this product

Catalog No. 19-038-942

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) For assembly, grinding, machine operations, metal production and more
    Color Black
    Quantity 10 Case
    Product Type Knob
    For Use With (Equipment) 3M™ Headgear L-Series

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit