Learn More
3M™ Lexa™ GoggleGear™
Wider view than traditional impact goggles
Manufacturer:  3M™ 166150000010
 View more versions of this product
Catalog No. 19-038-484A
  Check Availability
Add to cart
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings and limitations accompanying each product.
Specifications Description & Specifications
Specifications
Safety Goggles
Black
Foam backing seals, Top and bottom vent channels, Adjustable Temple Frame
Medium
Antifog; Hardcoat
ANSI Z87.1-2010-High Impact; CSA Z94.3-2007 standards
Black
Clear
Antifog, hardcoat