3M™ Lexa™ Safety Eyewear
No need for browguards or sideshields
Manufacturer: 3M™ 152120000020
 View more versions of this product
Catalog No. 19-807-981
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance Meets the requirements of ANSI Z87.1-2010 and CSA Z94.3-2007.
    Lens Material Polycarbonate
    Lens Tint Green Shade
    Lens Coating Anti-fog; Hardcoated
    Temple Color Black
    Combine extra-wide viewing range and contoured design that follows facial outline; eliminates the need for bulky browguards and distracting sideshields.
    • Lightweight, full-coverage eye protection
    • DX™ anti-fog hard coat protects lens from fog, scratching, and most chemicals
    • Lens are easy to switch — simply snap off the temples and re-attach them to the desired lens
    • Fully customizable with three lens angles and four temple lengths to fit virtually everyone
    • Slate temples
  • Description & Specifications
Related Products
Product Content Feedback