Learn More
3M™ Lexa Safety Glasses
Lexa fighter protective eyewear combines extra wide viewing range and contoured design, and is fully customizable with multiple lens options, three lens angles and four temple lengths. Lightweight, high wrap offers additional coverage.
Catalog Number | Mfr. No. | Lens Tint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Lens Tint | Price | Quantity & Availability | |||||
19-807-977
|
3M™
7010045714 |
Clear | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
19-038-483B
|
3M™
152140000020 |
Indoor/Outdoor Mirror | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | View substitute products » |
Description
These safety glasses are recommended for the following tasks: assembly, chipping, chiseling, cleaning, drilling, grinding, machining, masonry, metal repair, painting pouring/casting, power fastening, riveting, sanding, and sawing. Industries in which these safety glasses are commonly used include aviation, construction, food and beverage, general manufacturing, mining, oil and gas, primary metals, and transportation. 3M Catalog# 15152-00000-100Specifications
Anti-fog |