Learn More
3M™ Maxim™ 2X2 Safety Goggles
Versatile design for tacticle use
Manufacturer:  3M™ 7000052838
 View more versions of this product
Catalog No. 19-060-0423
Add to cart
Description
Description
Lightweight safety goggle was designed to help protect from sun, wind, dust and flying debris.
  • Dual-lens design enhances sightline and field of vision
  • Vented air seal designed to help minimize fogging
  • Featuring flexible fit temples
  • Interchangeable temples and broad elastic strap
  • 3M Catalog# 40686-00000-10 , 3M Catalog# 40687-00000-10
Compliance
Meets the High Impact Requirements of ANSI Z87.1-2003.
Specifications
Specifications
Safety goggles
Safety
ANSI Z87.1-2003
Black
Clear
10/Cs.
Provide protection against hazards such as dust and chemical splash