Learn More
3M™ Maxim™ Eyewear
Contemporary, athletic styling
Manufacturer:  3M™ 132530000020
 View more versions of this product
Catalog No. 19-121-578
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
Description
Feauture polycarbonate lenses that absorb 99.9% UV.
  • Three-position pantascopic lens angle adjustment
  • Dual-injected cushioned browguard
  • Soft nose adjustable nose bridge
  • Adjustable temple lengths
  • Ventilation channel helps to prevent fogging
Compliance
Meets ANSI Z87.1-2015 standards.
Specifications
Specifications
Meets ANSI Z87.1-2010 and CSA Z94.3-2007 standards.
Black
Amber
Polycarbonate
Adjustable
Anti-fog