Learn More
3M™ Metaliks™ Eyewear
Catalog Number 19121271
Click to view available options
Frame Color:
Bronze
Brushed Nickel
Polished Metal
Silver
Lens Coating:
Anti-fog
Lens Tint:
Blue Mirror
Bronze
Clear
Gray
Indoor/Outdoor Mirror
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
This item is not returnable. View return policy
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Offers distinctive style, maximum coverage, and adjustable comfort
  • Impact-resistant polycarbonate lenses with antifog hardcoat† to resist fog, scratches, and most chemicals
  • Soft, adjustable nose bridge and temple tips
    • Standard Metaliks feature a 10-base de-centered lens for optimum front and side protection
    • Metaliks Sports feature brushed nickel frame with athletic-shaped lens design
    • Metaliks GT feature a race-inspired design with a bronze frame
    • 3M Catalog# 11343-10000-20, 11344-10000-20, 11345-10000-20, 11554-00000-20, 11555-00000-20, 11556-00000-20, 15170-10000-20, 15172-10000-20
Compliance
Meets the requirements of ANSI™ Z87.1-2010 and CSA Z94.3-2007.
†I/O Mirror lens have hardcoat only, not antifog hardcoat.
Lens Coating Anti-fog
Certifications/Compliance Meets the requirements of ANSI Z87.1-2010 and CSA Z94.3-2007.
Lens Tint Clear
Frame Color Brushed Nickel
Model 11343-10000-20