Learn More
3M™ Metaliks™ Eyewear
Offers distinctive style, maximum coverage, and adjustable comfort
Supplier: 3M™ 7010315203
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Description
- Impact-resistant polycarbonate lenses with antifog hardcoat† to resist fog, scratches, and most chemicals
- Soft, adjustable nose bridge and temple tips
- Standard Metaliks feature a 10-base de-centered lens for optimum front and side protection
- Metaliks Sports feature brushed nickel frame with athletic-shaped lens design
- Metaliks GT feature a race-inspired design with a bronze frame
- 3M Catalog# 11343-10000-20, 11344-10000-20, 11345-10000-20, 11554-00000-20, 11555-00000-20, 11556-00000-20, 15170-10000-20, 15172-10000-20
†I/O Mirror lens have hardcoat only, not antifog hardcoat.
Compliance
Meets the requirements of ANSI™ Z87.1-2010 and CSA Z94.3-2007.
Specifications
Meets the requirements of ANSI Z87.1-2010 and CSA Z94.3-2007. | |
Bronze | |
11555-00000-20 |
Anti-fog | |
Bronze |