Learn More
3M™ Metaliks™ Eyewear
Offers distinctive style, maximum coverage, and adjustable comfort
Supplier:  3M™ 7010385323
 View more versions of this product
Catalog No. 19-810-522
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
This item is not returnable. View return policy
Description
Description
  • Impact-resistant polycarbonate lenses with antifog hardcoat† to resist fog, scratches, and most chemicals
  • Soft, adjustable nose bridge and temple tips
    • Standard Metaliks feature a 10-base de-centered lens for optimum front and side protection
    • Metaliks Sports feature brushed nickel frame with athletic-shaped lens design
    • Metaliks GT feature a race-inspired design with a bronze frame
    • 3M Catalog# 11343-10000-20, 11344-10000-20, 11345-10000-20, 11554-00000-20, 11555-00000-20, 11556-00000-20, 15170-10000-20, 15172-10000-20
    †I/O Mirror lens have hardcoat only, not antifog hardcoat.
    Compliance
    Meets the requirements of ANSI™ Z87.1-2010 and CSA Z94.3-2007.
Specifications
Specifications
Meets the requirements of ANSI Z87.1-2010 and CSA Z94.3-2007.
Blue Mirror
11556-00000-20
Hardcoat
Bronze
Hardcoated