Learn More
3M™ Nassau™ Plus™ Eyewear
Click to view available options
Frame Color:
Black
Red, White, Blue
Description
- Lightweight but durable
- Frame features adjustable temples for a custom fit
- Integrated sideshield for wraparound protection
- Single-piece high-impact polycarbonate lens
- Comfortable flared nosepiece with high set bridge option ideal for broad or low nasal profile
- DX™ antifog hard coat protects lens from fog, scratching, and most chemicals
- 3M Catalog# 14300-00000-20 , 3M Catalog# 14327-00000-20
Compliance
Meets ANSI Z87.1-2003 and CSA Z94.3-2007 standards
Specifications
Certifications/Compliance | ANSI Z87.1-2003, CSA Z94.1-2007 |
Lens Tint | Clear |
Frame Color | Black |
Model | 14300-00000-20 |
Lens Material | Polycarbonate |