Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ COMTAC™ III Advanced Combat Helmet

Third generation of COMTAC Tactical Communication Headsets designed to meet the mission needs of today's warfighter

Specifications

Certifications/Compliance MIL-STD-810F, EMI Standard MIL-STD-461E
Color Foliage Green
For Use With (Equipment) PRR radio
View More Specs

Includes: Gel cushions, PTT, carrying bag and batteries

Products 1
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
19-153-658
View Documents Promotion Details
3M™
MT17H682FB19PRR
Dual comm head band, ambidextrous noise-canceling boom mic, external microphones, rechargeable or disposable 2 AAA battery (500+ hours battery life), hardwired downlead cable coming off the back of each cup with one lead This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Level-dependent surround sound for “talk-thru” function
  • External communications and stereo talk-thru are independent of each other
  • Ambidextrous noise-canceling boom mic
  • Gel cushion option
  • Adjustable sizing to fit different users
  • Available with single comm or dual comm headband
  • Color: foliage green

Compliance

Environmental Standard MIL-STD-810F; EMI Standard MIL-STD-461E

Specifications

Specifications

MIL-STD-810F, EMI Standard MIL-STD-461E
Foliage Green
PRR radio
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit