Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Peltor™ ComTac™ III Advanced Combat Helmet Kit

The COMTAC III ACH headset is the third generation of COMTAC Tactical Communication Headsets designed to meet the mission needs of today's warfighter.

Specifications

Certifications/Compliance MIL-STD-810F, EMI Standard MIL-STD-461E
Color Foliage Green
For Use With (Equipment) AN/PRC-148, AN/PRC-152, AN/PRC-117, AN/PRC-119 radios
View More Specs

Includes: Gel cushions, PTT, carrying bag and batteries

Products 3
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
19-153-662
View Documents Promotion Details
3M™
7010341814
Gel cushions, one PTT, carrying bag, batteries; Foliage green This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-153-661
View Documents Promotion Details
3M™
7010384188
Gel cushion, two PTT, carrying bag, batteries This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
19-153-663
View Documents Promotion Details
3M™
7010384189
Gel cushion, dual comm PTT, bag, battery; Foliage green This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

  • Level-dependent surround sound for talk-thru function
  • External communications and stereo talk-thru are independent of each other
  • Ambidextrous noise-canceling boom mic
  • Gel cushion option
  • Adjustable sizing to fit different users
  • Color: foliage green
  • 3M Catalog# 88061-00000 , 3M Catalog# 88062-00000 , 3M Catalog# 88064-00000

For radios AN/PRC-148, AN/PRC-152, AN/PRC-117, AN/PRC-119

Compliance

Environmental Standard MIL-STD-810F; EMI Standard MIL-STD-461E

Specifications

Specifications

MIL-STD-810F, EMI Standard MIL-STD-461E
Foliage Green
AN/PRC-148, AN/PRC-152, AN/PRC-117, AN/PRC-119 radios
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit