3M™ Peltor™ ComTac™ III Advanced Combat Helmet Kit

The COMTAC III ACH headset is the third generation of COMTAC Tactical Communication Headsets designed to meet the mission needs of today's warfighter.

Manufacturer: 3M™ 8806100000

 View more versions of this product

Catalog No. 19-153-661

  Check Availability
Add to Cart

    Specifications

    Color Foliage Green
    Certifications/Compliance MIL-STD-810F, EMI Standard MIL-STD-461E
    Includes Gel cushion, two PTT, carrying bag, batteries
    For Use With (Equipment) AN/PRC-148, AN/PRC-152, AN/PRC-117, AN/PRC-119 radios

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit