COMTAC™ III Advanced Combat Helmet

Third generation of COMTAC Tactical Communication Headsets designed to meet the mission needs of today's warfighter

Manufacturer: 3M MT17H682FB47PRR

 View more versions of this product

Catalog No. 19-153-657

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) PRR radio

    • Level-dependent surround sound for “talk-thru” function
    • External communications and stereo talk-thru are independent of each other
    • Ambidextrous noise-canceling boom mic
    • Gel cushion option
    • Adjustable sizing to fit different users
    • Available with single comm or dual comm headband
    • Color: foliage green

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit