Learn More
3M™ COMTAC™ III Advanced Combat Helmet
Third generation of COMTAC Tactical Communication Headsets designed to meet the mission needs of today's warfighter
Manufacturer:  3M™ MT17H682FB19PRR
 View more versions of this product
Catalog No. 19-153-658
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
MIL-STD-810F, EMI Standard MIL-STD-461E
Foliage Green
PRR radio
Dual comm head band, ambidextrous noise-canceling boom mic, external microphones, rechargeable or disposable 2 AAA battery (500+ hours battery life), hardwired downlead cable coming off the back of each cup with one lead