Learn More
3M™ PELTOR™ FLX2 Cable
Consists of an assortment of PELTOR headsets and cables that allow for a simple and reliable connection to a wide variety of handheld devices. Flexible and adaptable, the FLX2 platform allows you to pair your protective communication needs with your existing handheld devices.
Manufacturer:  3M™ 7100219827
 View more versions of this product
Catalog No. 03-051-110
Add to cart
Description
Description
  • FLX2 plug for connection to a wide assortment PELTOR headsets
  • Kevlar™ reinforced
  • Robust and highly flexible
  • Polyurethane cable
  • Open ended cable allows for customization
Specifications
Specifications
FLX2-210
No
No
Straight Open End
Non Pertinent
60 /Cs
10 m
Polyurethane, Kevlar™ Reinforced
No