3M™ Peltor™ HearPlugs™ Listen-Only Earpiece

Combines high-quality audio reception with NVLAP certified noise reduction

Manufacturer: 3M™ PELTIP1

 View more versions of this product

Catalog No. 19-121-320

  Check Availability
Add to Cart

Alert

The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.


    Specifications

    Product Type Peltor™ Hearplug Listen Earpiece
    Hearing Protection Style In ear
    Mission Type Aircraft, ships, vehicles
    Noise Reduction Range (NRR) 21/29dB
    Certifications/Compliance CSA Class AL
    Color Yellow/black
    For Use With (Application) Designed for two-way radio users who require clear reception of incoming transmission in high-noise areas

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit