Learn More
3M™ Peltor™ NightHawk™ Tactical Goggle Kit
DX™ coating resists fog, scratching, and many chemicals
Manufacturer:  3M™ 4059400000
Catalog No. 19-169-511
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.
Includes: MOLLE-attachable protective case
Description
Description
Built specifically to provide protection from sand, dirt, and debris.
  • Full 180° view for better peripheral vision
  • Designed to provide protection from sand, dirt, and debris
  • Interchangeable goggles and spectacles
  • Night vision compatible
  • Innovative top and bottom vent system designed to resist powdery sand while helping to minimize fogging
  • Lightweight, microfiber lens shroud helps prevent scratching and keeps lens clean
  • Soft polymer seal flexes to fit virtually every face shape snuggly and comfortably around the eyes, forehead, and nose without gapping or pinching
  • Headstrap slides and clips on Fit Loop so goggles fit easily under or over helmets
Order Info
Universal prescription insert available for Tactical Goggle Kit; contact your Fisher Safety Customer Service Representative for details.
Compliance
Goggle and spectacle meet Military Vo Regimen for high velocity impact, high impact requirements of ANSI Z87.1-2003, Buy American Act and Berry Amendment.