3M™ Peltor™ PTL™ Ear Muffs

Allow workers to listen safely—no need for removal

Manufacturer: 3M™ H7APTL

 View more versions of this product

Catalog No. 19-130-1975

  Check Availability
Add to Cart

Alert

The 3M products in this catalog must be used in accordance with OSHA regulations, and the user instructions, warnings, and limitations accompanying each product.


    Specifications

    Certifications/Compliance UL
    Color Black
    Cushion Material Liquid-foam
    Dielectric No
    For Use With (Application) Assembly, cleaning, demolition, facility maintenance, grinding, machine operations, painting, sanding, welding
    Includes Padded headband
    Material Foam Plastic Metal
    Noise Reduction Rating (NRR) 26dB
    Style Over-the-head
    Product Type Ear Muff

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit