Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Powerflow™ Face-Mounted Powered Air Purifying Respirators

Powered APR provides protection against dust, mist, fumes, radionuclides and radon daughters. Ideal for lead and asbestos.

Supplier:  3M™ 6900PF

 View more versions of this product

Catalog No. 19-822-577

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes:
  • Medium full facepiece
  • Motor blower unit
  • Battery pack
  • Flow meter

Description

Description

  • High efficiency particulate filter
  • Delivers minimum of 4cfm when used with filter
  • NiCd rechargeable eight-hour battery
  • System uses 6000 Series facepiece
  • Faceshield meets High Impact Requirements of ANSI Z87.1-2003
  • Complete system includes series 6000 full facepiece, 6000DIN, motor blower unit, battery pack, and flow indicator
Specifications

Specifications

ANSI Z87.1-2003
PAPR System with Facepiece
4 CFM
Medium full facepiece, motor blower unit, battery pack, flow meter
Face-mounted
2.131 kg
HE Filter
Large
Lead, asbestos, and hazardous particulate
6900PF
4.7 lb.
SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.